Assay Details
| Gene Symbol: RnrL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057679.5 | NP_477027.1 | 3 - 4 | 387 | 104 | |
| GenBank mRNA | AF132143.1 | - | 3 - 4 | 370 | 104 | |
| AY119149.1 | - | 3 - 4 | 430 | 104 | ||
More Information
Set Membership: |
|

RnrL
Genomic Map