Assay Details
| Gene Symbol: ana | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299297.1 | NP_001286226.1 | 2 - 3 | 566 | 73 | |
| NM_057690.3 | NP_477038.1 | 2 - 3 | 911 | 73 | ||
| GenBank mRNA | BT132678.1 | - | 2 - 3 | 903 | 73 | |
More Information
Set Membership: |
|

ana
Genomic Map