Assay Details
| Gene Symbol: ana | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169622.2 | NP_001163093.1 | 3 - 4 | 1167 | 61 | |
| NM_001299297.1 | NP_001286226.1 | 3 - 4 | 813 | 61 | ||
| NM_057690.3 | NP_477038.1 | 3 - 4 | 1158 | 61 | ||
| GenBank mRNA | BT132678.1 | - | 3 - 4 | 1150 | 61 | |
More Information
Set Membership: |
|

ana
Genomic Map