Assay Details
| Gene Symbol: ana | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169622.2 | NP_001163093.1 | 4 - 5 | 1241 | 67 | |
| NM_001299297.1 | NP_001286226.1 | 4 - 5 | 887 | 67 | ||
| NM_057690.3 | NP_477038.1 | 4 - 5 | 1232 | 67 | ||
| GenBank mRNA | BT132678.1 | - | 4 - 5 | 1224 | 67 | |
More Information
Set Membership: |
|

ana
Genomic Map