Assay Details
| Gene Symbol: Pen | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057693.5 | NP_477041.1 | 4 - 5 | 1329 | 71 | |
| GenBank mRNA | BT003258.1 | - | 4 - 5 | 1239 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pen| Gene Symbol: Pen | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057693.5 | NP_477041.1 | 4 - 5 | 1329 | 71 | |
| GenBank mRNA | BT003258.1 | - | 4 - 5 | 1239 | 71 | |
Set Membership: |
|