Assay Details
| Gene Symbol: Ack-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274005.1 | NP_001260934.1 | 5 - 6 | 2339 | 73 | |
| NM_001274006.1 | NP_001260935.1 | 5 - 6 | 2339 | 73 | ||
| NM_057738.4 | NP_477086.2 | 5 - 6 | 2339 | 73 | ||
| NM_057739.4 | NP_477087.3 | 5 - 6 | 2339 | 73 | ||
| GenBank mRNA | AY051755.1 | - | 5 - 6 | 2318 | 73 | |
More Information
Set Membership: |
|

Ack-like
Genomic Map