Assay Details
| Gene Symbol: Ack-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057738.4 | NP_477086.2 | 6 - 7 | 2562 | 62 | |
| GenBank mRNA | AY051755.1 | - | 6 - 7 | 2541 | 62 | |
More Information
Set Membership: |
|

Ack-like
Genomic Map