Assay Details
| Gene Symbol: Ack-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057738.4 | NP_477086.2 | 7 - 8 | 3880 | 68 | |
| NM_057739.4 | NP_477087.3 | 7 - 8 | 3883 | 68 | ||
| GenBank mRNA | AY051755.1 | - | 7 - 8 | 3859 | 68 | |
More Information
Set Membership: |
|

Ack-like
Genomic Map