Assay Details
| Gene Symbol: Rab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273803.1 | NP_001260732.1 | 1 - 2 | 192 | 70 | |
| NM_057742.4 | NP_477090.1 | 1 - 2 | 192 | 70 | ||
| GenBank mRNA | BT003315.1 | - | 1 - 2 | 146 | 70 | |
| D84313.1 | - | 1 - 2 | 144 | 70 | ||
More Information
Set Membership: |
|

Rab2
Genomic Map