Assay Details
| Gene Symbol: Diap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057779.5 | NP_477127.1 | 1 - 2 | 521 | 80 | |
| NM_176182.2 | NP_788362.1 | 2 - 3 | 412 | 80 | ||
| GenBank mRNA | AY051844.1 | - | 1 - 2 | 490 | 80 | |
More Information
Set Membership: |
|

Diap2
Genomic Map