Assay Details
| Gene Symbol: Nap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259556.1 | NP_001246485.1 | 3 - 4 | 694 | 74 | |
| NM_001259557.2 | NP_001246486.1 | 3 - 4 | 677 | 74 | ||
| NM_057780.4 | NP_477128.1 | 3 - 4 | 662 | 74 | ||
| GenBank mRNA | BT001479.1 | - | 3 - 4 | 609 | 74 | |
| BT120094.1 | - | 3 - 4 | 694 | 74 | ||
More Information
Set Membership: |
|

Nap1
Genomic Map