Assay Details
Gene Symbol: Nap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259556.1 | NP_001246485.1 | 5 - 6 | 1027 | 74 | |
NM_001259557.2 | NP_001246486.1 | 5 - 6 | 1010 | 74 | ||
NM_057780.4 | NP_477128.1 | 5 - 6 | 995 | 74 | ||
GenBank mRNA | BT001479.1 | - | 5 - 6 | 942 | 74 | |
BT120094.1 | - | 5 - 6 | 1027 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
