Assay Details
| Gene Symbol: csw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272245.1 | NP_001259174.1 | 2 - 3 | 901 | 64 | |
| NM_057782.5 | NP_477130.1 | 2 - 3 | 379 | 64 | ||
| GenBank mRNA | BT001484.1 | - | 2 - 3 | 313 | 64 | |
| M94730.1 | - | 2 - 3 | 348 | 64 | ||
More Information
Set Membership: |
|

csw
Genomic Map