Assay Details
| Gene Symbol: Orc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057784.8 | NP_477132.1 | 1 - 2 | 1298 | 61 | |
| GenBank mRNA | AY089590.1 | - | 1 - 2 | 1279 | 61 | |
| U43505.1 | - | 1 - 2 | 1098 | 61 | ||
More Information
Set Membership: |
|

Orc5
Genomic Map