Assay Details
| Gene Symbol: dare | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057802.6 | NP_477150.1 | 2 - 3 | 736 | 65 | |
| GenBank mRNA | BT083432.1 | - | 2 - 3 | 733 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dare| Gene Symbol: dare | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057802.6 | NP_477150.1 | 2 - 3 | 736 | 65 | |
| GenBank mRNA | BT083432.1 | - | 2 - 3 | 733 | 65 | |
Set Membership: |
|