Assay Details
| Gene Symbol: Clp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057808.2 | NP_477156.1 | 1 | 1120 | 114 | |
| GenBank mRNA | BT025009.1 | - | 1 | 1112 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clp| Gene Symbol: Clp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057808.2 | NP_477156.1 | 1 | 1120 | 114 | |
| GenBank mRNA | BT025009.1 | - | 1 | 1112 | 114 | |
Set Membership: |
|