Assay Details
| Gene Symbol: nrv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273234.1 | NP_001260163.1 | 2 - 3 | 843 | 71 | |
| NM_057819.4 | NP_477167.1 | 2 - 3 | 843 | 71 | ||
| GenBank mRNA | BT044185.1 | - | 2 - 3 | 813 | 71 | |
More Information
Set Membership: |
|

nrv1
Genomic Map