Assay Details
| Gene Symbol: nrv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014475.2 | NP_001014475.1 | 1 - 2 | 443 | 64 | |
| NM_001273235.1 | NP_001260164.1 | 2 - 3 | 935 | 64 | ||
| NM_057820.4 | NP_477168.1 | 3 - 4 | 968 | 64 | ||
| GenBank mRNA | AY060289.1 | - | 1 - 2 | 430 | 64 | |
More Information
Set Membership: |
|

nrv2
Genomic Map