Assay Details
| Gene Symbol: Acer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273329.1 | NP_001260258.1 | 2 - 3 | 961 | 73 | |
| NM_057847.4 | NP_477195.1 | 2 - 3 | 961 | 73 | ||
| GenBank mRNA | AY051750.1 | - | 2 - 3 | 766 | 73 | |
More Information
Set Membership: |
|

Acer
Genomic Map