Assay Details
Gene Symbol: Pkg21D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057865.4 | NP_477213.1 | 2 - 3 | 679 | 102 | |
GenBank mRNA | AY058288.1 | - | 2 - 3 | 662 | 102 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
