Assay Details
| Gene Symbol: Pkg21D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057865.4 | NP_477213.1 | 4 - 5 | 2318 | 62 | |
| GenBank mRNA | AY058288.1 | - | 4 - 5 | 2301 | 62 | |
More Information
Set Membership: |
|

Pkg21D
Genomic Map