Assay Details
| Gene Symbol: Rca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057866.3 | NP_477214.1 | 1 | 1327 | 151 | |
| GenBank mRNA | AY058372.1 | - | 1 | 1301 | 151 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rca1| Gene Symbol: Rca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057866.3 | NP_477214.1 | 1 | 1327 | 151 | |
| GenBank mRNA | AY058372.1 | - | 1 | 1301 | 151 | |
Set Membership: |
|