Assay Details
| Gene Symbol: Cdk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057878.4 | NP_477226.2 | 3 - 4 | 663 | 75 | |
| GenBank mRNA | AF027300.1 | - | 3 - 4 | 640 | 75 | |
| AY118809.1 | - | 3 - 4 | 645 | 75 | ||
More Information
Set Membership: |
|

Cdk9
Genomic Map