Assay Details
| Gene Symbol: Lcp65Ac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057931.5 | NP_477279.1 | 1 - 2 | 219 | 102 | |
| GenBank mRNA | AY071303.1 | - | 1 - 2 | 214 | 102 | |
| KX532141.1 | - | 1 - 2 | 127 | 102 | ||
| U84755.1 | - | 1 - 2 | 202 | 102 | ||
More Information
Set Membership: |
|

Lcp65Ac
Genomic Map