Assay Details
| Gene Symbol: Ac13E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014745.2 | NP_001014745.1 | 2 - 3 | 479 | 68 | |
| NM_001272646.2 | NP_001259575.1 | 3 - 4 | 1234 | 68 | ||
| NM_057951.3 | NP_477299.1 | 2 - 3 | 479 | 68 | ||
More Information
Set Membership: |
|

Ac13E
Genomic Map