Assay Details
| Gene Symbol: Uba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273902.1 | NP_001260831.1 | 1 - 2 | 1048 | 65 | |
| NM_057962.5 | NP_477310.2 | 1 - 2 | 1048 | 65 | ||
| GenBank mRNA | AY069191.1 | - | 1 - 2 | 1048 | 65 | |
More Information
Set Membership: |
|

Uba1
Genomic Map