Assay Details
| Gene Symbol: Orc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057972.4 | NP_477320.1 | 1 - 2 | 317 | 93 | |
| GenBank mRNA | BT004868.1 | - | 1 - 2 | 261 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Orc4| Gene Symbol: Orc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057972.4 | NP_477320.1 | 1 - 2 | 317 | 93 | |
| GenBank mRNA | BT004868.1 | - | 1 - 2 | 261 | 93 | |
Set Membership: |
|