Assay Details
| Gene Symbol: Parg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258582.2 | NP_001245511.1 | 1 - 2 | 835 | 98 | |
| NM_057973.4 | NP_477321.2 | 1 - 2 | 849 | 98 | ||
| GenBank mRNA | AY051955.1 | - | 1 - 2 | 841 | 98 | |
More Information
Set Membership: |
|

Parg
Genomic Map