Assay Details
| Gene Symbol: ebi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057981.4 | NP_477329.1 | 1 - 2 | 378 | 83 | |
| GenBank mRNA | BT133073.1 | - | 1 - 2 | 343 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ebi| Gene Symbol: ebi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057981.4 | NP_477329.1 | 1 - 2 | 378 | 83 | |
| GenBank mRNA | BT133073.1 | - | 1 - 2 | 343 | 83 | |
Set Membership: |
|