Assay Details
| Gene Symbol: Srp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057999.5 | NP_477347.1 | 1 | 1749 | 84 | |
| GenBank mRNA | AY069568.1 | - | 1 | 1713 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srp54| Gene Symbol: Srp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057999.5 | NP_477347.1 | 1 | 1749 | 84 | |
| GenBank mRNA | AY069568.1 | - | 1 | 1713 | 84 | |
Set Membership: |
|