Assay Details
| Gene Symbol: mys | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298068.1 | NP_001284997.1 | 6 - 7 | 2678 | 56 | |
| NM_001298069.1 | NP_001284998.1 | 6 - 7 | 2823 | 56 | ||
| NM_001298070.1 | NP_001284999.1 | 6 - 7 | 3033 | 56 | ||
| NM_080054.3 | NP_524793.2 | 6 - 7 | 2678 | 56 | ||
| GenBank mRNA | AY113499.1 | - | 6 - 7 | 2676 | 56 | |
More Information
Set Membership: |
|

mys
Genomic Map