Assay Details
| Gene Symbol: Spn43Ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032224.2 | NP_001027395.1 | 3 - 4 | 1131 | 60 | |
| GenBank mRNA | BT030771.1 | - | 3 - 4 | 1172 | 60 | |
More Information
Set Membership: |
|

Spn43Ab
Genomic Map