Assay Details
| Gene Symbol: eEF1beta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299577.1 | NP_001286506.1 | 1 - 2 | 171 | 59 | |
| NM_080069.4 | NP_524808.3 | 1 - 2 | 171 | 59 | ||
More Information
Set Membership: |
|

eEF1beta
Genomic Map