Assay Details
| Gene Symbol: cv-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080070.4 | NP_524809.2 | 5 - 6 | 1631 | 63 | |
| GenBank mRNA | BT133425.1 | - | 5 - 6 | 1629 | 63 | |
More Information
Set Membership: |
|

cv-2
Genomic Map