Assay Details
| Gene Symbol: sens | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080079.3 | NP_524818.1 | 1 - 2 | 376 | 58 | |
| GenBank mRNA | AF212312.1 | - | 1 - 2 | 377 | 58 | |
| BT022164.1 | - | 1 - 2 | 368 | 58 | ||
More Information
Set Membership: |
|

sens
Genomic Map