Assay Details
| Gene Symbol: Abl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275030.1 | NP_001261959.1 | 1 - 2 | 1071 | 75 | |
| NM_001275032.1 | NP_001261961.1 | 1 - 2 | 1071 | 75 | ||
| NM_001275035.1 | NP_001261964.1 | 1 - 2 | 1071 | 75 | ||
| NM_001300156.1 | NP_001287085.1 | 1 - 2 | 1071 | 75 | ||
| NM_080104.3 | NP_524843.2 | 1 - 2 | 1071 | 75 | ||
More Information
Set Membership: |
|

Abl
Genomic Map