Assay Details
| Gene Symbol: Abl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104153.3 | NP_001097623.1 | 8 - 9 | 3161 | 77 | |
| NM_001275034.1 | NP_001261963.1 | 8 - 9 | 3161 | 77 | ||
| NM_001275035.1 | NP_001261964.1 | 7 - 8 | 3107 | 77 | ||
| NM_001275036.1 | NP_001261965.1 | 8 - 9 | 3161 | 77 | ||
| NM_001300156.1 | NP_001287085.1 | 7 - 8 | 3107 | 77 | ||
| NM_080104.3 | NP_524843.2 | 7 - 8 | 3107 | 77 | ||
More Information
Set Membership: |
|

Abl
Genomic Map