Assay Details
| Gene Symbol: ImpE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274648.1 | NP_001261577.1 | 4 - 5 | 805 | 66 | |
| NM_080110.3 | NP_524849.2 | 4 - 5 | 749 | 66 | ||
More Information
Set Membership: |
|

ImpE1
Genomic Map