Assay Details
| Gene Symbol: S6KL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258813.1 | NP_001245742.1 | 7 - 8 | 951 | 61 | |
| NM_001258814.2 | NP_001245743.1 | 7 - 8 | 1423 | 61 | ||
| NM_080122.4 | NP_524861.2 | 7 - 8 | 1429 | 61 | ||
| GenBank mRNA | AY069491.1 | - | 7 - 8 | 1323 | 61 | |
More Information
Set Membership: |
|

S6KL
Genomic Map