Assay Details
| Gene Symbol: Lk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080154.5 | NP_524893.2 | 1 | 568 | 128 | |
| GenBank mRNA | BT023388.1 | - | 1 | 562 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lk| Gene Symbol: Lk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080154.5 | NP_524893.2 | 1 | 568 | 128 | |
| GenBank mRNA | BT023388.1 | - | 1 | 562 | 128 | |
Set Membership: |
|