Assay Details
| Gene Symbol: fs(2)ltoPP43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273673.1 | NP_001260602.1 | 1 - 2 | 313 | 70 | |
| NM_080201.3 | NP_524940.1 | 1 - 2 | 313 | 70 | ||
| NM_176062.2 | NP_788076.1 | 1 - 2 | 313 | 70 | ||
More Information
Set Membership: |
|

fs(2)ltoPP43
Genomic Map