Assay Details
| Gene Symbol: fs(2)ltoPP43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080201.3 | NP_524940.1 | 2 - 3 | 774 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fs(2)ltoPP43| Gene Symbol: fs(2)ltoPP43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080201.3 | NP_524940.1 | 2 - 3 | 774 | 79 | |
Set Membership: |
|