Assay Details
| Gene Symbol: Spn42Dd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169596.3 | NP_001163067.1 | 2 - 3 | 906 | 103 | |
| GenBank mRNA | BT010110.1 | - | 2 - 3 | 878 | 103 | |
More Information
Set Membership: |
|

Spn42Dd
Genomic Map