Assay Details
| Gene Symbol: geko | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080241.3 | NP_524980.1 | 2 - 3 | 820 | 62 | |
| GenBank mRNA | AB008048.1 | - | 2 - 3 | 819 | 62 | |
| AY071271.1 | - | 2 - 3 | 818 | 62 | ||
More Information
Set Membership: |
|

geko
Genomic Map