Assay Details
Gene Symbol: wds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258574.2 | NP_001245503.1 | 2 - 3 | 1196 | 91 | |
NM_080245.5 | NP_524984.1 | 2 - 3 | 1196 | 91 | ||
GenBank mRNA | AY118386.1 | - | 2 - 3 | 1193 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
