Assay Details
Gene Symbol: Vhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273956.1 | NP_001260885.1 | 1 | 743 | 101 | |
NM_080247.3 | NP_524986.1 | 1 | 743 | 101 | ||
GenBank mRNA | AF195836.1 | - | 1 | 721 | 101 | |
AY075555.1 | - | 1 | 742 | 101 | ||
BT023867.1 | - | 1406 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
