Assay Details
| Gene Symbol: dbo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274963.1 | NP_001261892.1 | 3 - 4 | 1032 | 51 | |
| NM_080250.3 | NP_524989.2 | 3 - 4 | 1032 | 51 | ||
| GenBank mRNA | AB027612.1 | - | 3 - 4 | 1026 | 51 | |
| BT010272.1 | - | 3 - 4 | 1028 | 51 | ||
More Information
Set Membership: |
|

dbo
Genomic Map