Assay Details
| Gene Symbol: cmet | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273451.1 | NP_001260380.1 | 6 - 7 | 5546 | 83 | |
| NM_001273452.1 | NP_001260381.1 | 7 - 8 | 5711 | 83 | ||
| NM_001273453.1 | NP_001260382.1 | 6 - 7 | 5546 | 83 | ||
| NM_080254.3 | NP_524993.3 | 7 - 8 | 5711 | 83 | ||
More Information
Set Membership: |
|

cmet
Genomic Map