Assay Details
| Gene Symbol: mRpL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272219.1 | NP_001259148.1 | 1 - 2 | 139 | 66 | |
| NM_080302.3 | NP_525041.1 | 1 - 2 | 139 | 66 | ||
| GenBank mRNA | AY058507.1 | - | 1 - 2 | 96 | 66 | |
More Information
Set Membership: |
|

mRpL16
Genomic Map