Assay Details
| Gene Symbol: pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272253.1 | NP_001259182.1 | 5 - 6 | 6291 | 89 | |
| NM_001272254.2 | NP_001259183.2 | 5 - 6 | 6303 | 89 | ||
| NM_080306.4 | NP_525045.3 | 5 - 6 | 6291 | 89 | ||
More Information
Set Membership: |
|

pcx
Genomic Map